偏苯三酸三辛酯 ≥97.0% (by GC)
供应商: TCI
Synonyms:
偏苯三酸三(2-乙基己基)酯, TOTM
B0881-25ML
B0881-500ML
TCIAB0881-25MLEA
0
CNY
TCIAB0881-25ML
TCIAB0881-500ML
偏苯三酸三辛酯 ≥97.0% (by GC)
偏苯三酸三辛酯
分子式:
C₃₃H₅₄O₆ |
MDL编号:
MFCD00053799 CAS号: 3319-31-1 EINECS: 222-020-0 |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...