10-十一碳炔酸 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Undec-10-ynoic acid
U0054-5G
U0054-1G
TCIAU0054-5GEA
0
CNY
TCIAU0054-5G
TCIAU0054-1G
10-十一碳炔酸 ≥98.0% (by GC)
10-十一碳炔酸
分子式:
C₁₁H₁₈O₂ 分子量: 182.26 g/mol 沸点: 180°C |
MDL编号:
MFCD00014389 CAS号: 2777-65-3 EINECS: 220-471-8 |
|
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...