2-萘甲醇 ≥98.0% (by GC)

供应商: TCI

N0881-25G N0881-5G
TCIAN0881-25GEA 0 CNY
TCIAN0881-25G TCIAN0881-5G
2-萘甲醇 ≥98.0% (by GC)
2-萘甲醇
分子式: C₁₁H₁₀O
沸点: 178°C
MDL编号: MFCD00004124
CAS号: 1592-38-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db5d1dac53c4bc08c3e537a88c20c724461be84f&vid=db5d1dac53c4bc08c3e537a88c20c724461be84f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR