2-萘甲醇 98%
供应商: Thermo Fisher Scientific
351150010
351150050
ACRO351150010EA
0
CNY
ACRO351150010
ACRO351150050
2-萘甲醇 98%
2-萘甲醇
分子式:
C₁₁H₁₀O 分子量: 158.2000 g/mol 溶点: 79…83 °C 存储条件: Ambient |
MDL编号:
MFCD00004124 CAS号: 1592-38-7 |
产品技术规格
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 79 to 83 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...