1-萘甲醇 98%

供应商: Thermo Fisher Scientific

Synonyms: Naphthalen-1-ylmethanol

180110020 180110100
ACRO180110020EA 0 CNY
ACRO180110020 ACRO180110100
1-萘甲醇 98%
1-萘甲醇
分子式: C₁₁H₁₀O
分子量: 158.20 g/mol
沸点: 301 °C (715 mmHg)
溶点: 59…62 °C
存储条件: Ambient
MDL编号: MFCD00004044
CAS号: 4780-79-4
EINECS: 225-324-1

现在订购

产品技术规格

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 59 to 62 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR