1-萘乙醇 ≥95.0% (by GC)

供应商: TCI

Synonyms: 2-(1-萘基)乙醇

N0575-25G N0575-5G
TCIAN0575-25GEA 0 CNY
TCIAN0575-25G TCIAN0575-5G
1-萘乙醇 ≥95.0% (by GC)
1-萘乙醇
分子式: C₁₂H₁₂O
沸点: 179°C
MDL编号: MFCD00004050
CAS号: 773-99-9
EINECS: 212-260-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2e7e33075b4562f77f331e71a883a13a12b5b18&vid=a2e7e33075b4562f77f331e71a883a13a12b5b18&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 59.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR