1-萘基乙酰胺 ≥99.0% (by GC)

供应商: TCI

N0050-25G N0050-250G
TCIAN0050-25GEA 0 CNY
TCIAN0050-25G TCIAN0050-250G
1-萘基乙酰胺 ≥99.0% (by GC)
1-萘基乙酰胺
分子式: C₁₂H₁₁NO
MDL编号: MFCD00059128
CAS号: 575-36-0
EINECS: 209-380-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b622bda7386eb363b35b54af879e44edc265e43&vid=2b622bda7386eb363b35b54af879e44edc265e43&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 159.0 to 162.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR