Choline bromide ≥98.0% (通过滴定分析)
供应商: TCI
|
Warning
|
C0328-25G
C0328-500G
TCIAC0328-25GEA
0
CNY
TCIAC0328-25G
TCIAC0328-500G
Choline bromide ≥98.0% (通过滴定分析)
Choline bromide
分子式:
C₅H₁₄BrNO 溶点: 298°C |
MDL编号:
MFCD00054244 CAS号: 1927-06-6 EINECS: 217-660-2 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...