4'-Bromobenzo-15-crown-5 (4'-bromobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥93.0% (by GC)

供应商: TCI
Warning

Synonyms: 4'-Bromobenzo-15-crown 5-Ether, 2,3-(4-Bromobenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene, 15-Bromo-2,3,5,6,8,9,11,12-octahydrobenzo[b][1,4,7,10,13]pentaoxacyclopentadecine

B2189-1G B2189-5G
TCIAB2189-1GEA 0 CNY
TCIAB2189-1G TCIAB2189-5G
4'-Bromobenzo-15-crown-5 (4'-bromobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥93.0% (by GC)
4'-Bromobenzo-15-crown-5 (4'-bromobenzo-1,4,7,10,13-pentaoxacyclopentadecane)
分子式: C₁₄H₁₉BrO₅
分子量: 347.2000 g/mol
MDL编号: MFCD00143270
CAS号: 60835-72-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9aa8c8da44845dc356cd73ed66f294c6b7b1f3e2&vid=9aa8c8da44845dc356cd73ed66f294c6b7b1f3e2&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 93.0 %
Melting point 63.0 to 70.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR