1,1-二苯乙烯 ≥98.0% (通过GC) stabilised
供应商: TCI
Stabilization:
Stabilised with hydroquinone (HQ)
D0886-25G
D0886-250G
D0886-5G
TCIAD0886-25GEA
0
CNY
TCIAD0886-25G
TCIAD0886-250G
TCIAD0886-5G
1,1-二苯乙烯 ≥98.0% (通过GC) stabilised
1,1-二苯乙烯
分子式:
(C₆H₅)₂CCH₂ 沸点: 270 °C (1013 hPa) 密度: 1,03 g/cm³ (20 °C) 存储条件: Refrigerator |
MDL编号:
MFCD00008583 CAS号: 530-48-3 EINECS: 208-482-6 Merck Index: 14,03323 |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...