1-氯-3-甲基-2-丁烯 ≥85.0% (by GC) stabilised
供应商: TCI
Danger
Stabilization: Stabilised with potassium carbonate
C1290-25ML C1290-250ML
TCIAC1290-25MLEA 0 CNY
TCIAC1290-25ML TCIAC1290-250ML
1-氯-3-甲基-2-丁烯 ≥85.0% (by GC) stabilised
1-氯-3-甲基-2-丁烯
分子式: C₅H₉Cl
分子量: 104.58 g/mol
沸点: 109°C
闪点: 15°C
存储条件: Refrigerator
MDL编号: MFCD00000982
CAS号: 503-60-6
EINECS: 207-972-7
UN号: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=912c01e8301d0d33a6a775cf5f02ce6536ab5b6b&vid=06942cca8bfa03000debde50f91a196f3e96c17d&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 85.0 %