1-氟-3-碘-5-硝基苯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
F0815-5G
F0815-1G
TCIAF0815-5GEA
0
CNY
TCIAF0815-5G
TCIAF0815-1G
1-氟-3-碘-5-硝基苯 ≥98.0% (by GC)
1-氟-3-碘-5-硝基苯
分子式:
C₆H₃FINO₂ 分子量: 267 g/mol |
MDL编号:
MFCD00007219 CAS号: 3819-88-3 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...