Methyl-4-amino-3-methylbenzoate ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: Methyl 4-amino-3-methylbenzoate

M2152-25G M2152-5G
TCIAM2152-25GEA 0 CNY
TCIAM2152-25G TCIAM2152-5G
Methyl-4-amino-3-methylbenzoate ≥98.0% (by GC, titration analysis)
Methyl-4-amino-3-methylbenzoate
分子式: C₉H₁₁NO₂
分子量: 165.19 g/mol
MDL编号: MFCD00102230
CAS号: 18595-14-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=260edfe26687462247ddc0dbeca7d618fa02cb44&vid=260edfe26687462247ddc0dbeca7d618fa02cb44&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 118.0 to 122.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR