3-氨基-2-甲基苯甲酸甲酯 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: Methyl 3-amino-2-methylbenzoate, 3-Amino-o-toluic acid Methyl Ester, 3-Amino-2-methylbenzoic acid Methyl Ester

A1563-25G A1563-5G
TCIAA1563-25GEA 0 CNY
TCIAA1563-25G TCIAA1563-5G
3-氨基-2-甲基苯甲酸甲酯 ≥98.0% (by GC, titration analysis)
3-氨基-2-甲基苯甲酸甲酯
分子式: C₉H₁₁NO₂
分子量: 165.19 g/mol
沸点: 160°C
闪点: 104°C
MDL编号: MFCD00211328
CAS号: 18583-89-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbaf427caef4fee82c1debace76b1be8d98cdca8&vid=bbaf427caef4fee82c1debace76b1be8d98cdca8&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR