4-Isobutylbenzoic acid ≥99.0% (通过GC,滴定分析)

供应商: TCI
Warning

I0437-10G I0437-25G
TCIAI0437-10GEA 0 CNY
TCIAI0437-10G TCIAI0437-25G
4-Isobutylbenzoic acid ≥99.0% (通过GC,滴定分析)
4-Isobutylbenzoic acid
分子式: C₁₁H₁₄O₂
MDL编号: MFCD00191658
CAS号: 38861-88-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39c215739ac51c60dab800d078a86c6d52c48d84&vid=39c215739ac51c60dab800d078a86c6d52c48d84&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 143.0 to 146.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR