5-(4-甲基苯基)-1H-四唑 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 5-(p-Tolyl)-1H-tetrazole

T2486-25G T2486-5G
TCIAT2486-25GEA 0 CNY
TCIAT2486-25G TCIAT2486-5G
5-(4-甲基苯基)-1H-四唑 ≥98.0% (by HPLC, titration analysis)
5-(4-甲基苯基)-1H-四唑
分子式: C₈H₈N₄
分子量: 160.18 g/mol
溶点: 248°C
MDL编号: MFCD01318164
CAS号: 24994-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70208244916a01e12b65bdf64953d8d0f3677e8b&vid=70208244916a01e12b65bdf64953d8d0f3677e8b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR