Ethyl-2-ethylbutyrate ≥99.0% (通过GC)

供应商: TCI
Warning

E0345-25ML E0345-500ML
TCIAE0345-25MLEA 0 CNY
TCIAE0345-25ML TCIAE0345-500ML
Ethyl-2-ethylbutyrate ≥99.0% (通过GC)
Ethyl-2-ethylbutyrate
分子式: C₈H₁₆O₂
沸点: 152°C
闪点: 39°C
MDL编号: MFCD00059396
CAS号: 2983-38-2
EINECS: 221-044-9
UN号: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=137c14207e1a8824fc41e8895c18335da687d346&vid=137c14207e1a8824fc41e8895c18335da687d346&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR