5,6,7,8-四氢喹喔啉 ≥98.0% (通过GC,滴定分析)

供应商: TCI
Warning

T1403-25G T1403-5G
TCIAT1403-25GEA 0 CNY
TCIAT1403-25G TCIAT1403-5G
5,6,7,8-四氢喹喔啉 ≥98.0% (通过GC,滴定分析)
5,6,7,8-四氢喹喔啉
分子式: C₈H₁₀N₂
闪点: 90°C
MDL编号: MFCD00038078
CAS号: 34413-35-9
EINECS: 252-002-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=321920e539a1fc9e993476d5c69242a742ff0754&vid=321920e539a1fc9e993476d5c69242a742ff0754&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR