5-甲基-3-氨基-2-氯吡啶 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-Chloro-5-methylpyridin-3-amine
A2405-25G
A2405-5G
TCIAA2405-25GEA
0
CNY
TCIAA2405-25G
TCIAA2405-5G
5-甲基-3-氨基-2-氯吡啶 ≥98.0% (by GC)
5-甲基-3-氨基-2-氯吡啶
分子式:
C₆H₇ClN₂ 分子量: 142.5900 g/mol |
MDL编号:
MFCD03427656 CAS号: 34552-13-1 |
|
产品技术规格
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 87.0 to 91.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...