4-甲氧基苯基溴化镁 1 M in THF, AcroSeal®

供应商: Thermo Fisher Scientific
Danger

Synonyms: 4-Anisylmagnesium bromide

377421000 377428000
ACRO377421000EA 0 CNY
ACRO377421000 ACRO377428000
4-甲氧基苯基溴化镁 1 M in THF, AcroSeal®
4-甲氧基苯基溴化镁
分子式: C₇H₇BrMgO
分子量: 211.34 g/mol
密度: 1 g/cm³
闪点: –21 °C
存储条件: Ambient
MDL编号: MFCD00013515
CAS号: 13139-86-1
UN号: 2924
ADR: 3 II

现在订购

产品技术规格

Appearance Yellow to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 0.9 to 1.2 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR