Methyl-4-benzyloxybenzoate ≥98.0% (通过GC)

供应商: TCI

M2180-25G M2180-5G
TCIAM2180-25GEA 0 CNY
TCIAM2180-25G TCIAM2180-5G
Methyl-4-benzyloxybenzoate ≥98.0% (通过GC)
分子式: C₁₅H₁₄O₃
MDL编号: MFCD00017613
CAS号: 32122-11-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e1256252024ed487fb180cd3587ae514a935a49&vid=7e1256252024ed487fb180cd3587ae514a935a49&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR