Sulphamethoxypyridazine ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Danger

Synonyms: 4-Amino-N-(6-methoxypyridazin-3-yl)benzenesulphonamide

S0591-25G S0591-5G
TCIAS0591-25GEA 0 CNY
TCIAS0591-25G TCIAS0591-5G
Sulphamethoxypyridazine ≥98.0% (by HPLC, titration analysis)
Sulphamethoxypyridazine
分子式: C₁₁H₁₂N₄O₃S
分子量: 280.3 g/mol
MDL编号: MFCD00057372
CAS号: 80-35-3
EINECS: 201-272-5
Merck Index: 14,08919
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c853b34eb7c6bad768d2c1c8d67348e9e7ef9fa&vid=0c853b34eb7c6bad768d2c1c8d67348e9e7ef9fa&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 180.0 to 184.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR