α-溴-4-氯苯乙酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Danger

B3367-25G B3367-5G
TCIAB3367-25GEA 0 CNY
TCIAB3367-25G TCIAB3367-5G
α-溴-4-氯苯乙酸 ≥98.0% (by titrimetric analysis)
α-溴-4-氯苯乙酸
分子式: C₈H₆BrClO₂
分子量: 249.49 g/mol
MDL编号: MFCD08276760
CAS号: 3381-73-5
UN号: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d2b46f02d46f1d637a491fa59fa1f6daafa09e0&vid=1d2b46f02d46f1d637a491fa59fa1f6daafa09e0&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR