4-噁唑甲酸乙酯 ≥96.0% (by GC)

供应商: TCI

Synonyms: 噁唑-4-羧酸乙酯

O0289-1G O0289-5G
TCIAO0289-1GEA 0 CNY
TCIAO0289-1G TCIAO0289-5G
4-噁唑甲酸乙酯 ≥96.0% (by GC)
4-噁唑甲酸乙酯
分子式: C₆H₇NO₃
分子量: 141.1300 g/mol
沸点: 101°C
溶点: 48°C
存储条件: Refrigerator
MDL编号: MFCD04114940
CAS号: 23012-14-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR