4,5-二氯-3-羟基哒嗪 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 4,5-二氯哒嗪-3(2H)-酮

D2768-100G D2768-25G
TCIAD2768-100GEA 0 CNY
TCIAD2768-100G TCIAD2768-25G
4,5-二氯-3-羟基哒嗪 ≥98.0% (by GC, titration analysis)
4,5-二氯-3-羟基哒嗪
分子式: C₄H₂Cl₂N₂O
分子量: 164.98 g/mol
MDL编号: MFCD00051504
CAS号: 932-22-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30c703c41a9b4ddfd9619dd36f82443a89abac75&vid=30c703c41a9b4ddfd9619dd36f82443a89abac75&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 201.0 to 206.0 deg-C
Solubility in hot EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR