4-氯甲基-3,5-二甲基异噁唑 ≥95.0% (by GC)
供应商: TCI
|
Warning
|
C2282-1G
C2282-5G
TCIAC2282-1GEA
0
CNY
TCIAC2282-1G
TCIAC2282-5G
4-氯甲基-3,5-二甲基异噁唑 ≥95.0% (by GC)
4-氯甲基-3,5-二甲基异噁唑
分子式:
C₆H₈ClNO 沸点: 88°C 闪点: 96°C 存储条件: Refrigerator |
MDL编号:
MFCD00003154 CAS号: 19788-37-5 EINECS: 243-312-4 |
|
产品技术规格
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...