四氟硼酸卓 ≥98.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: 四氟硼酸卓鎓

T1121-5G T1121-1G
TCIAT1121-5GEA 0 CNY
TCIAT1121-5G TCIAT1121-1G
四氟硼酸卓 ≥98.0% (by titrimetric analysis)
四氟硼酸卓
分子式: C₇H₇BF₄
存储条件: Refrigerator
MDL编号: MFCD00013402
CAS号: 27081-10-3
EINECS: 248-214-5
UN号: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cb04a9974aee54c5309fea992ecff8ba496752e&vid=2cb04a9974aee54c5309fea992ecff8ba496752e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR