3-Ethyl-2-methylbenzoxazolium iodide ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

E0477-25G E0477-5G
TCIAE0477-25GEA 0 CNY
TCIAE0477-25G TCIAE0477-5G
3-Ethyl-2-methylbenzoxazolium iodide ≥98.0% (by HPLC, titration analysis)
3-Ethyl-2-methylbenzoxazolium iodide
分子式: C₁₀H₁₂INO
MDL编号: MFCD00044141
CAS号: 5260-37-7
EINECS: 226-068-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b0b02b21698468733c359df7c2681216493f5df&vid=0b0b02b21698468733c359df7c2681216493f5df&prid=web&clid=SAPDAM

现在订购

产品技术规格

Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR