Filter plates, 96-well, for solvent filtration, AcroPrep™ Advance
供应商: CYTIVA PALL
8047 8048 8247 8148 8147 8248
PALL8047EA 0 CNY
PALL8047 PALL8048 PALL8247 PALL8148 PALL8147 PALL8248
Filter plates, 96-well, for solvent filtration, AcroPrep™ Advance
微孔板 过滤板
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes that provide the best possible performance for a wide range of sample preparation and high throughput screening applications. Available with a choice of PTFE or PTFE membrane.
  • Chemically resistant materials provide a stable platform to process samples in organic solvents
  • Low, non specific binding: inert construction materials ensure complete sample recovery
  • Well geometry results in faster, more uniform filtration rates across the plate with reduced hold-up volume
  • Output tip geometry allows direct flow of samples into receiver plate reducing cross-contamination risk
The PTFE (hydrophilic polytetrafluoroethene) membrane is an all-purpose, hydrophilic membrane for aqueous, acidic, basic, non-aggressive organic, and aggressive organic solutions.
The wwPTFE (hydrophillic polytetrafluoroethene) membrane is an all-purpose, hydrophilic membrane for aqueous, acidic, basic, non-aggressive organic, and aggressive organic solutions. The wwPTFE membrane offers low protein binding and low levels of UV-absorbing extractables.
Dimensions
L×W×H (with lid): 128×86×18 mm (350 μl only)
L×W×H (without lid): 128×86×18 mm (350 μl); 128×86×33 mm (1 ml); 128×86×47 mm (2 ml)
Well bottom area: 0,25 cm²
Recommended operating vacuum: ≥254 mm Hg
Recommended centrifugal force: 1500×g
Materials of construction
Filter media: PTFE (polytetrafluoroethylene) membrane or wwPTFE membrane (water wettable polytetrafluoroethylene) membrane
Plate housing: PP
Lid: PS