Culture tubes, reusable, with screw cap, KIMBLE®
供应商: KIMBLEKONTES
45066-20150 45066-38200 45066-25200
KIML45066-20150EA 0 CNY
KIML45066-20150 KIML45066-38200 KIML45066-25200
Culture tubes, reusable, with screw cap, KIMBLE®
管子 培养管
Manufactured from 33 expansion, low extractable borosilicate glass, KIMAX®.
Tubes with uniform wall thickness for maximum heat transfer and chemical resistance. Tubes feature a marking spot and a threaded phenolic screw cap that can be loosened for autoclaving. Tubes are available with rubber-lined caps or PTFE-faced rubber-lined caps.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements. Tubes with rubber-lined caps are designed from ASTM Specification E982, Type VI, Class A requirements. Tubes with PTFE-faced rubber-lined caps are designed from ASTM Specification E982, Type VI, Class B requirements.
订购须知 Cap is supplied unattached.