Sample vials, with PTFE lined screw cap, KIMBLE®
供应商: KIMBLEKONTES
60940A-8 60940A-4
KIML60940A-8EA 0 CNY
KIML60940A-8 KIML60940A-4
Sample vials, with PTFE lined screw cap, KIMBLE®
小瓶
Sample vials made from clear KIMAX® KG-33 borosilicate glass.
  • Closed top white PTFE closures, attached
  • The 16 ml size complies with EPA 600 Series Methods