Chemical resistant gloves, Alto 415
供应商: MAPA
34415040 34415151 34415046 34415047 34415048 34415049
MAPA34415040EA 0 CNY
MAPA34415040 MAPA34415151 MAPA34415046 MAPA34415047 MAPA34415048 MAPA34415049
Chemical resistant gloves, Alto 415
手套
These neoprene/natural latex gloves with textured finish and cotton flock lining are suitable for spray-painting, industrial cleaning, maintenance and for work with composite materials in the aeronautics industry.
  • Tactile sensitivity, due to the gloves reduced thickness
  • Cotton flock-lining esnures comfort and perspiration absorption
  • Good grip thanks to the non-slip embossing
Certifications: EN 388:2016 (1011X), EN ISO 374-1 Type B (KMT), EN ISO 374-5, EN 421; Cat. III