Chemical resistant gloves, FluoTech 344
供应商: MAPA
34344130 34344139
MAPA34344130EA 0 CNY
MAPA34344130 MAPA34344139
Chemical resistant gloves, FluoTech 344
手套
These fluoroelastomer gloves with textile lining are recommended for prolonged contact with aromatic solvents, sulphur-based chemicals and amines.
  • Excellent comfort for long tasks, due to the cotton knit interior
  • High mechanical resistance and long service life, due to textile support
  • Freedom of movement: good glove flexibility
Ideal for use in the chemical industry, for metal degreasing and etching, site pollution control and petrochemical refining.
Certifications: EN 388:2016 (3121X), EN ISO 374-1 Type A (ACDEFGJLMN), EN ISO 374-5, EN 407 (X1XXXX); Cat. III
Caution: Fluoroelastomer is not recommended for prolonged contact with ketones, organic acids and acetates.