Multi-channel pipettes, electronic, variable volume, VOYAGER
供应商: INTEGRA
4722 4724 4721 4723 4731 4736
INTL4722EA 0 CNY
INTL4722 INTL4724 INTL4721 INTL4723 INTL4731 INTL4736
Multi-channel pipettes, electronic, variable volume, VOYAGER
移液管和移液器 移液器
These electronic pipettes with motorised tip spacing enable parallel transfers of multiple samples between labware of different sizes.
  • Adjustable tip spacing provides access to a wide range of labware formats such as microplates, tube racks or gel boxes
  • GripTips seal effortlessly to optimise accuracy and precision
  • Low force tip load and ejection to reduce arm and wrist related stresses
  • Colour screen and touch wheel interface offer quick and comprehensive menu navigation
  • Set of ten predefined pipetting modes; personalised pipetting protocols can be created
Tip spacing can be changed by the simple push of a button, no manual adjustments or two handed operations are needed. This increases pipetting productivity and reduces the risk of developing a repetitive strain injury (RSI). Ideal for a wide variety of genomic, proteomic and cell culture applications.
订购须知 Units are supplied with rechargeable battery (located inside the pipette, Li-ion, 3,7 V), a bag of spare O-rings (300 µl and 1250 µl volume ranges only), O-ring removal tool (300 µl and 1250 µl volume ranges only) and accuracy and precision certificate. Supplied without power supply and tips. Those need to be ordered separately.