Ethyl-2,3,4,5-tetrafluorobenzoate ≥98.0% (通过GC)

供应商: TCI

T2094-100G T2094-25G
TCIAT2094-100GEA 0 CNY
TCIAT2094-100G TCIAT2094-25G
Ethyl-2,3,4,5-tetrafluorobenzoate ≥98.0% (通过GC)
Ethyl-2,3,4,5-tetrafluorobenzoate
分子式: C₉H₆F₄O₂
沸点: 88°C
MDL编号: MFCD02094158
CAS号: 122894-73-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=240c0ac7acdaf4901d4c809bea53a40c426563ca&vid=240c0ac7acdaf4901d4c809bea53a40c426563ca&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR