2-甲基-3-硝基苯甲酸甲酯 ≥98.0% (通过GC)

供应商: TCI

M2144-25G M2144-5G
TCIAM2144-25GEA 0 CNY
TCIAM2144-25G TCIAM2144-5G
2-甲基-3-硝基苯甲酸甲酯 ≥98.0% (通过GC)
2-甲基-3-硝基苯甲酸甲酯
分子式: C₉H₉NO₄
MDL编号: MFCD00082564
CAS号: 59382-59-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22b98218b11719b1bfefb6272cfea792e1c72136&vid=22b98218b11719b1bfefb6272cfea792e1c72136&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR