tert-Butylhydrazine hydrochloride ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

B1195-25G B1195-500G
TCIAB1195-25GEA 0 CNY
TCIAB1195-25G TCIAB1195-500G
tert-Butylhydrazine hydrochloride ≥98.0% (by titrimetric analysis)
tert-Butylhydrazine hydrochloride
分子式: C₄H₁₃ClN₂
分子量: 124.61 g/mol
MDL编号: MFCD00012947
CAS号: 7400-27-3
EINECS: 231-001-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09dc84fbe88d1ec17d78154d2dfc8ae0264febd9&vid=09dc84fbe88d1ec17d78154d2dfc8ae0264febd9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Purity (Potassium iodate Method) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR