(S)-(-)-2-Chlorobutyric acid ≥98.0% (通过滴定分析)
供应商: TCI
|
Danger
|
C1377-1G
C1377-5G
TCIAC1377-1GEA
0
CNY
TCIAC1377-1G
TCIAC1377-5G
(S)-(-)-2-Chlorobutyric acid ≥98.0% (通过滴定分析)
(S)-(-)-2-Chlorobutyric acid
分子式:
C₄H₇ClO₂ 沸点: 94°C |
MDL编号:
MFCD00067099 CAS号: 32653-32-0 UN号: 3265 ADR: 8,III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 94.0 % |
Optical purity (GC) | min. 96.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -13.0 to -19.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...