2-十一烷基咪唑 ≥96.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
2-Undecyl-1H-imidazole
U0012-25G
U0012-500G
TCIAU0012-25GEA
0
CNY
TCIAU0012-25G
TCIAU0012-500G
2-十一烷基咪唑 ≥96.0% (by titrimetric analysis)
2-十一烷基咪唑
分子式:
C₁₄H₂₆N₂ 分子量: 222.3696 g/mol |
MDL编号:
MFCD00038694 CAS号: 16731-68-3 EINECS: 240-796-9 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 93.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 71.0 to 75.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...