苄基缩水甘油醚 ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-((Benzyloxy)methyl)oxirane

B2108-25G B2108-5G
TCIAB2108-25GEA 0 CNY
TCIAB2108-25G TCIAB2108-5G
苄基缩水甘油醚 ≥97.0% (by GC)
苄基缩水甘油醚
分子式: C₁₀H₁₂O₂
存储条件: Refrigerator
MDL编号: MFCD00068664
CAS号: 2930-05-4
EINECS: 220-899-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3075e6d87bf9c2ff9c7cdb1e2f1c0e18693656e0&vid=3075e6d87bf9c2ff9c7cdb1e2f1c0e18693656e0&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR