1,3-Dimethyl-4-piperidone ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 1,3-Dimethylpiperidin-4-one

D3909-25G D3909-5G
TCIAD3909-25GEA 0 CNY
TCIAD3909-25G TCIAD3909-5G
1,3-Dimethyl-4-piperidone ≥98.0% (by GC, titration analysis)
1,3-Dimethyl-4-piperidone
分子式: C₇H₁₃NO
分子量: 127.1800 g/mol
沸点: 62°C
MDL编号: MFCD00100893
CAS号: 4629-80-5
EINECS: 225-046-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=618c11357e7c553cbe9abfa99b081ee45167286b&vid=618c11357e7c553cbe9abfa99b081ee45167286b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR