2-乙基己基丙烯酸酯 monomer ≥99.0% (by GC) stabilised
供应商: TCI
|
Warning
|
Synonyms:
(±)-丙烯酸-2-乙基己酯
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A0144-25ML
A0144-500ML
TCIAA0144-25MLEA
0
CNY
TCIAA0144-25ML
TCIAA0144-500ML
2-乙基己基丙烯酸酯 monomer ≥99.0% (by GC) stabilised
2-乙基己基丙烯酸酯
分子式:
C₁₁H₂₀O₂ 沸点: 215°C 溶点: –90°C 闪点: 80°C |
MDL编号:
MFCD00009495 CAS号: 103-11-7 EINECS: 203-080-7 |
|
产品技术规格
Appearance | Colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...