N-乙酰乙醇胺 ≥90.0% (by GC)

供应商: TCI
Danger

Synonyms: N-(2-羟基乙基)乙酰胺

A1935-25G A1935-500G
TCIAA1935-25GEA 0 CNY
TCIAA1935-25G TCIAA1935-500G
N-乙酰乙醇胺 ≥90.0% (by GC)
N-乙酰乙醇胺
分子式: C₄H₉NO₂
分子量: 103.12 g/mol
沸点: 167°C
闪点: 177°C
MDL编号: MFCD00002836
CAS号: 142-26-7
EINECS: 205-530-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae8952ae5c592e68cc0db98cacee51589bf2742d&vid=ae8952ae5c592e68cc0db98cacee51589bf2742d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR