1-Butyl-4-chlorobenzene ≥97.0% (通过GC)

供应商: TCI

B3143-25G B3143-5G
TCIAB3143-25GEA 0 CNY
TCIAB3143-25G TCIAB3143-5G
1-Butyl-4-chlorobenzene ≥97.0% (通过GC)
1-Butyl-4-chlorobenzene

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR