2,6-二甲氧基-4-羟基苯甲醛 ≥97.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 2,6-Dimethoxy-4-hydroxybenzaldehyde

H1326-5G H1326-1G
TCIAH1326-5GEA 0 CNY
TCIAH1326-5G TCIAH1326-1G
2,6-二甲氧基-4-羟基苯甲醛 ≥97.0% (by GC, titration analysis)
2,6-二甲氧基-4-羟基苯甲醛
分子式: C₉H₁₀O₄
MDL编号: MFCD01407645
CAS号: 22080-96-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bed319c4a21e1a9df0d3d42412f0e8d80f6cb5a&vid=3bed319c4a21e1a9df0d3d42412f0e8d80f6cb5a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR