6-Amino-1-naphthol ≥98.0% (通过HPLC)

供应商: TCI
Warning

A0360-1G A0360-5G
TCIAA0360-1GEA 0 CNY
TCIAA0360-1G TCIAA0360-5G
6-Amino-1-naphthol ≥98.0% (通过HPLC)
6-Amino-1-naphthol
分子式: C₁₀H₉NO
MDL编号: MFCD00059543
CAS号: 23894-12-4
EINECS: 245-933-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e091c531e2a90af598bf2424f936b8fcd0458cd4&vid=e091c531e2a90af598bf2424f936b8fcd0458cd4&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR