二乙基苯膦 ≥97.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: Diethyl(phenyl)phosphane

D1019-1ML D1019-5ML
TCIAD1019-1MLEA 0 CNY
TCIAD1019-1ML TCIAD1019-5ML
二乙基苯膦 ≥97.0% (by GC, titration analysis)
二乙基苯膦
分子式: C₁₀H₁₅P
沸点: 220°C
闪点: 85°C
MDL编号: MFCD00015172
CAS号: 1605-53-4
EINECS: 216-516-6
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b29c846a495d7f9f2952aecdba838be7916a72a9&vid=b29c846a495d7f9f2952aecdba838be7916a72a9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Iodometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR