D(+)-Talose ≥98.0% (by HPLC)

供应商: TCI

Synonyms: D-(+)-Talose, (2S,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal

T0869-100MG T0869-500MG
TCIAT0869-100MGEA 0 CNY
TCIAT0869-100MG TCIAT0869-500MG
D(+)-Talose ≥98.0% (by HPLC)
D(+)-Talose
分子式: C₆H₁₂O₆
MDL编号: MFCD00135834
CAS号: 2595-98-4
EINECS: 219-996-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6707027c35dfd02b898376718abe03910c66807e&vid=c2fff1d244b4a624bd35d19e463786340ba4838a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 132.0 to 136.0 deg-C
Specific rotation [a]20/D +20.0 to +23.0 deg (C=1, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR