D-阿洛糖 ≥98.0% (by HPLC)

供应商: TCI

Synonyms: β-D-阿洛糖, β-D-别吡喃糖, All

A1488-1G A1488-100MG
TCIAA1488-1GEA 0 CNY
TCIAA1488-1G TCIAA1488-100MG
D-阿洛糖 ≥98.0% (by HPLC)
D-阿洛糖
分子式: C₆H₁₂O₆
MDL编号: MFCD00135833
CAS号: 2595-97-3
EINECS: 219-994-4
Merck Index: 14,00280
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0269c5af634fe781c0996dac823b974f98a7093f&vid=0269c5af634fe781c0996dac823b974f98a7093f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +13.0 to +16.0 deg (C=5, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR