甲基丙烯酸-2-乙氧基乙酯 ≥98.0% (by GC) stabilised

供应商: TCI
Warning

Synonyms: Methacrylic acid 2-Ethoxyethyl ester, Ethylene glycol monoethyl ether Methacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M1251-25ML M1251-500ML
TCIAM1251-25MLEA 0 CNY
TCIAM1251-25ML TCIAM1251-500ML
甲基丙烯酸-2-乙氧基乙酯 ≥98.0% (by GC) stabilised
甲基丙烯酸-2-乙氧基乙酯
分子式: C₈H₁₄O₃
分子量: 158.1950 g/mol
沸点: 93°C
闪点: 72°C
MDL编号: MFCD00009249
CAS号: 2370-63-0
EINECS: 219-135-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=543352a1803af3bf7e8b4534d1bee3fb8a269249&vid=543352a1803af3bf7e8b4534d1bee3fb8a269249&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR